This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01110692_g1
See other CPSF6 GE Assays ›
Gene Symbol:
CPSF6
Gene Name:
cleavage and polyadenylation specific factor 6
Gene Aliases:
CFIM, CFIM68, HPBRII-4, HPBRII-7
Chromosome Location:
Chr.12: 69239537 - 69274358 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01110692_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CPSF6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300947.1 NP_001287876.1 6 - 7 921 82
XM_005268588.2 XP_005268645.1 6 - 7 921 82
GenBank mRNA BC000714.2 - 6 - 7 842 3506481 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA polyadenylation
mRNA processing
protein tetramerization
nucleotide binding
RNA binding
mRNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline