This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01111628_m1
See other PHTF2 GE Assays ›
Gene Symbol:
PHTF2
Gene Name:
putative homeodomain transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 77798792 - 77957504 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01111628_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHTF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127357.1 NP_001120829.1 3 - 4 239 149
NM_001127359.1 NP_001120831.1 3 - 4 239 149
XM_005250508.4 XP_005250565.1 4 - 5 291 149
XM_005250509.4 XP_005250566.1 4 - 5 216 149
XM_006716063.3 XP_006716126.1 4 - 5 291 149
XM_011516422.2 XP_011514724.1 4 - 5 291 149
XM_011516423.2 XP_011514725.1 4 - 5 291 149
GenBank mRNA BC022419.1 - 3 - 4 239 149
BC127093.1 - 3 - 4 155 40123852 149

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline