This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01111948_m1
See other HERC6 GE Assays ›
Gene Symbol:
HERC6
Gene Name:
HECT and RLD domain containing E3 ubiquitin protein ligase family member 6
Gene Aliases:
-
Chromosome Location:
Chr.4: 88378565 - 88443097 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01111948_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HERC6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165136.1 NP_001158608.1 19 - 20 2628 145
NM_017912.3 NP_060382.3 20 - 21 2736 145
XM_005263083.3 XP_005263140.1 20 - 21 2933 145
XM_011532053.2 XP_011530355.1 19 - 20 2827 145
XM_017008333.1 XP_016863822.1 19 - 20 2294 145
GenBank mRNA AF336798.1 - 20 - 21 2736 145
AK000644.1 - 7 - 8 824 145
AK097168.1 - 14 - 15 1551 145
AK225506.1 - 7 - 8 824 145
AK295832.1 - 20 - 21 2646 145
AL833664.1 - 9 - 10 1064 145
AY653201.1 - 19 - 20 2628 145
AY653202.1 - 19 - 20 2540 145
AY653203.1 - 17 - 18 2321 145
BC035775.1 - 6 - 7 676 145
BC042047.1 - 20 - 21 2612 5531519 145
BX647121.1 - 20 - 21 2412 145

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400008105.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000484.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HERC6

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
hematopoietic progenitor cell differentiation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 TC04000484.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 ENST00000264346.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 ENST00000380265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 ENST00000438983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 ENST00000511939.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 NM_001165136.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04007023.hg.1 NM_017912.3 Refseq
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000264346.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000273960.7 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000380265.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000506842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 NM_001165136.1 Refseq
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 NM_017912.3 Refseq
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000264346.11 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000273960.7 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000380265.9 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 ENST00000506842.1 Ensembl
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 NM_001165136.1 Refseq
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 NM_017912.3 Refseq
Affymetrix Human Clariom D Assay PSR0400155770.hg.1 TC0400008105.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0