This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01112184_m1
See other RIMS1 GE Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01112184_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIMS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168407.1 NP_001161879.1 16 - 17 2011 137
NM_001168408.1 NP_001161880.1 15 - 16 65
NM_001168409.1 NP_001161881.1 16 - 17 1799 137
NM_001168410.1 NP_001161882.1 16 - 17 2086 137
XM_005248688.3 XP_005248745.1 20 - 21 65
XM_011535604.2 XP_011533906.2 21 - 22 3534 137
XM_011535615.2 XP_011533917.2 20 - 21 65
XM_017010517.1 XP_016866006.1 21 - 22 3537 137
XM_017010518.1 XP_016866007.1 21 - 22 3534 137
XM_017010520.1 XP_016866009.1 20 - 21 65
XM_017010522.1 XP_016866011.1 21 - 22 3537 137
XM_017010523.1 XP_016866012.1 21 - 22 3537 137
XM_017010524.1 XP_016866013.1 20 - 21 65
XM_017010527.1 XP_016866016.1 20 - 21 65
XM_017010530.1 XP_016866019.1 21 - 22 3537 137
XM_017010531.1 XP_016866020.1 21 - 22 3534 137
XM_017010532.1 XP_016866021.1 20 - 21 65
XM_017010537.1 XP_016866026.1 15 - 16 65
XM_017010538.1 XP_016866027.1 16 - 17 2215 137
XM_017010539.1 XP_016866028.1 16 - 17 2102 137
XM_017010540.1 XP_016866029.1 15 - 16 65
XM_017010541.1 XP_016866030.1 15 - 16 65
XM_017010542.1 XP_016866031.1 15 - 16 65
XM_017010543.1 XP_016866032.1 15 - 16 65
XM_017010546.1 XP_016866035.1 15 - 16 65
XM_017010547.1 XP_016866036.1 15 - 16 65
XM_017010552.1 XP_016866041.1 16 - 17 2338 137
XM_017010553.1 XP_016866042.1 16 - 17 2335 137
XM_017010554.1 XP_016866043.1 15 - 16 65
XM_017010557.1 XP_016866046.1 3 - 4 65
XM_017010558.1 XP_016866047.1 3 - 4 230 137
GenBank mRNA AB051866.1 - 21 - 22 65
AF263305.1 - 9 - 10 65
AF263306.1 - 8 - 9 65
AF263309.1 - 8 - 9 65
AK293828.1 - 15 - 16 65
AK294868.1 - 16 - 17 2011 137
AK295001.1 - 15 - 16 65
AK296303.1 - 16 - 17 1855 137
AK309185.1 - 16 - 17 1799 137
AK316108.1 - 16 - 17 1812 137

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008505.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06000726.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017010557.1,XM_017010558.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 TC06000726.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000264839.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000348717.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000350827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000352008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000401910.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000491071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000517960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000521978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 ENST00000538414.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 NM_001168407.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008666.hg.1 NM_014989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 TC06000726.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 ENST00000401910.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 ENST00000425662.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 ENST00000517827.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 ENST00000521978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 ENST00000538414.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 NM_001168407.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 NM_001168409.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 NM_001168410.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008662.hg.1 NM_014989.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 TC06000726.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000264839.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000346609.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000348717.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000349908.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000350827.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000352008.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000401910.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000425662.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000491071.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000517827.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000517960.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000518273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000520567.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000521978.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000522291.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000523963.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 ENST00000538414.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 NM_001168407.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 NM_001168408.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 NM_001168409.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 NM_001168410.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06008664.hg.1 NM_014989.5 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000264839.11 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000370420.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000401910.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000425662.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000453976.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000463023.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000491071.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000517433.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000517827.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000517960.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000518273.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000520567.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000521978.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000522211.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000522291.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000523963.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168407.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168408.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168409.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168410.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_014989.5 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000264839.11 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000370420.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000401910.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000425662.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000453976.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000463023.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000491071.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000517433.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000517827.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000517960.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000518273.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000520567.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000521978.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000522211.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000522291.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 ENST00000523963.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168407.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168408.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168409.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_001168410.1 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 NM_014989.5 Refseq
Affymetrix Human Clariom D Assay PSR0600159119.hg.1 TC0600008505.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0