This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01112202_m1
See other RIMS1 GE Assays ›
Gene Symbol:
RIMS1
Gene Name:
regulating synaptic membrane exocytosis 1
Gene Aliases:
CORD7, RAB3IP2, RIM, RIM1
Chromosome Location:
Chr.6: 71886703 - 72403143 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01112202_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIMS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017010523.1 XP_016866012.1 24 - 25 3926 88
XM_017010538.1 XP_016866027.1 20 - 21 2634 88
XM_017010546.1 XP_016866035.1 18 - 19 2530 88
XM_017010547.1 XP_016866036.1 18 - 19 2527 88
GenBank mRNA AF263307.1 - 10 - 11 1179 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
intracellular protein transport
neurotransmitter secretion
visual perception
positive regulation of gene expression
glutamate secretion
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulated exocytosis
secretion
regulation of neurotransmitter secretion
regulation of catalytic activity
response to stimulus
long-term synaptic potentiation
membrane fusion
positive regulation of inhibitory postsynaptic potential
positive regulation of dendrite extension
positive regulation of excitatory postsynaptic potential
protein binding
SH3 domain binding
Rab GTPase binding
GTPase regulator activity
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0