This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01113210_m1
See other POLR3GL GE Assays ›
Gene Symbol:
POLR3GL
Gene Name:
RNA polymerase III subunit G like
Gene Aliases:
RPC32HOM, flj32422
Chromosome Location:
Chr.1: 145959037 - 145978848 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01113210_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3GL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032305.1 NP_115681.1 3 - 4 366 110
XM_005277431.4 XP_005277488.1 3 - 4 368 110
GenBank mRNA AK056984.1 - 3 - 4 354 110
BC004355.1 - 3 - 4 366 3357827 110
DQ418461.1 - 2 - 3 259 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100018282.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003108.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR3GL

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
positive regulation of type I interferon production
RNA polymerase III activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01048134.hg.1 TC01003108.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01048134.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01048134.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay PSR0100239227.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239227.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay PSR0100239227.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100239227.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay PSR0100239227.hg.1 TC0100018282.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369313.7 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000369314.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000471706.1 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 ENST00000622508.4 Ensembl
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq
Affymetrix Human Clariom D Assay TC0100018282.hg.1 NM_032305.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0