This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01113647_g1
See other GNPAT GE Assays ›
Gene Symbol:
GNPAT
Gene Name:
glyceronephosphate O-acyltransferase
Gene Aliases:
DAP-AT, DAPAT, DHAPAT, RCDP2
Chromosome Location:
Chr.1: 231241173 - 231277973 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01113647_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GNPAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001316350.1 NP_001303279.1 9 - 10 1539 69
NM_014236.3 NP_055051.1 10 - 11 1722 69
XM_005273313.4 XP_005273370.1 10 - 11 1682 69
XM_011544303.2 XP_011542605.1 11 - 12 1771 69
XM_011544304.2 XP_011542606.1 10 - 11 1590 69
GenBank mRNA AB209882.1 - 10 - 11 1627 69
AF035285.1 - 4 - 5 625 69
AF043937.1 - 10 - 11 1598 69
AJ002190.1 - 10 - 11 1673 69
AK222773.1 - 10 - 11 1678 69
AK223304.1 - 10 - 11 1681 69
AK297982.1 - 9 - 10 1500 69
AK311386.1 - 9 - 10 1372 69
AK314181.1 - 10 - 11 1685 69
BC000450.2 - 10 - 11 1569 2821393 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011958.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001915.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GNPAT

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerophospholipid metabolic process
phosphatidic acid biosynthetic process
synapse assembly
response to nutrient
ether lipid biosynthetic process
cerebellum morphogenesis
paranodal junction assembly
response to drug
response to starvation
membrane organization
response to fatty acid
receptor binding
glycerone-phosphate O-acyltransferase activity
palmitoyl-CoA hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01029793.hg.1 TC01001915.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01029793.hg.1 ENST00000366646.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029793.hg.1 ENST00000366647.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029793.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay PSR0100188234.hg.1 TC0100011958.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000366647.8 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000416000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000436239.5 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000469332.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 ENST00000492459.1 Ensembl
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq
Affymetrix Human Clariom D Assay TC0100011958.hg.1 NM_014236.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0