This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01114477_g1
See other ZP4 GE Assays ›
Gene Symbol:
ZP4
Gene Name:
zona pellucida glycoprotein 4
Gene Aliases:
ZBP, ZP1, ZPB, Zp-4
Chromosome Location:
Chr.1: 237877864 - 237890922 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01114477_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021186.4 NP_067009.1 8 - 9 1446 69
GenBank mRNA AK314151.1 - 8 - 9 1446 69
BC069521.1 - 8 - 9 1159 7262334 69
DV080723.1 - 1 - 2 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100017878.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01004010.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZP4

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of humoral immune response
binding of sperm to zona pellucida
protein kinase A signaling
intracellular signal transduction
positive regulation of T cell proliferation
positive regulation of protein kinase activity
acrosomal vesicle exocytosis
protein kinase C signaling
positive regulation of acrosome reaction
negative regulation of binding of sperm to zona pellucida
signal transducer activity
acrosin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01060706.hg.1 TC01004010.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01060706.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01060706.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay PSR0100234874.hg.1 TC0100017878.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000366570.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 ENST00000611898.4 Ensembl
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq
Affymetrix Human Clariom D Assay TC0100017878.hg.1 NM_021186.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline