This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01114992_m1
See other OGG1 GE Assays ›
Gene Symbol:
OGG1
Gene Name:
8-oxoguanine DNA glycosylase
Gene Aliases:
HMMH, HOGG1, MUTM, OGH1
Chromosome Location:
Chr.3: 9749944 - 9788246 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01114992_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016821.2 NP_058214.1 6 - 7 1295 97
GenBank mRNA U88620.1 - 6 - 7 1262 97
Y13277.1 - 6 - 7 1256 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300006556.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000042.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OGG1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
base-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
response to radiation
response to light stimulus
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
response to drug
negative regulation of apoptotic process
depurination
depyrimidination
response to ethanol
response to folic acid
cellular response to cadmium ion
negative regulation of double-strand break repair via single-strand annealing
damaged DNA binding
endonuclease activity
protein binding
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
oxidized purine DNA binding
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 TC03000042.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 ENST00000302008.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 ENST00000302036.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 ENST00000349503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 NM_016821.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 NM_016826.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 NM_016827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 NM_016828.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000857.hg.1 NM_016829.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 TC03000042.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 ENST00000302008.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 ENST00000302036.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 ENST00000349503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 ENST00000383826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 ENST00000449570.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 NM_016821.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 NM_016826.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 NM_016827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 NM_016828.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000855.hg.1 NM_016829.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 TC03000042.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 ENST00000302008.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 ENST00000302036.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 ENST00000349503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 ENST00000449570.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 NM_016821.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 NM_016826.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 NM_016827.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 NM_016828.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03000856.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000302008.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000302036.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000349503.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000352937.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000383826.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000449570.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016821.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016826.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016827.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016828.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000302008.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000302036.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000349503.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000352937.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000383826.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 ENST00000449570.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016821.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016826.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016827.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016828.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145210.hg.1 TC0300006556.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000302008.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000302036.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000349503.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000352937.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016821.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016826.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016827.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016828.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000302008.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000302036.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000349503.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 ENST00000352937.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016821.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016826.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016827.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016828.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145213.hg.1 TC0300006556.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000302008.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000302036.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000349503.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000352937.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000449570.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016821.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016826.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016827.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016828.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000302008.12 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000302036.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000349503.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000352937.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 ENST00000449570.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016821.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016826.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016827.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016828.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 NM_016829.2 Refseq
Affymetrix Human Clariom D Assay PSR0300145212.hg.1 TC0300006556.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0