This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115000_m1
See other AADAC GE Assays ›
Gene Symbol:
AADAC
Gene Name:
arylacetamide deacetylase
Gene Aliases:
CES5A1, DAC
Chromosome Location:
Chr.3: 151814008 - 151828488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115000_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AADAC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001086.2 NP_001077.2 3 - 4 524 139
XM_005247104.4 XP_005247161.1 3 - 4 538 139
GenBank mRNA AK290628.1 - 3 - 4 475 139
BC032309.1 - 3 - 4 459 5185099 139
L32179.1 - 3 - 4 519 139

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300009192.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000830.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AADAC

Panther Classification:

Molecular Function -

deacetylase

Gene Ontology Categories:

Function(s) Process(es)

xenobiotic metabolic process
positive regulation of triglyceride catabolic process
catalytic activity
triglyceride lipase activity
lipase activity
serine hydrolase activity
deacetylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000462937.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000462937.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000462937.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000496128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000496128.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03014825.hg.1 TC03000830.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03014825.hg.1 ENST00000232892.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03014825.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03014825.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009192.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000462937.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000462937.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000462937.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000463420.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000471766.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000483636.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000496128.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000496128.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 ENST00000561502.5 Ensembl
Affymetrix Human Clariom D Assay TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Clariom D Assay TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Clariom D Assay TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Clariom D Assay TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Clariom D Assay TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Clariom D Assay TC0300009190.hg.1 NR_026915.1 Refseq
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 ENST00000232892.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 ENST00000488869.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 NM_001086.2 Refseq
Affymetrix Human Clariom D Assay PSR0300165995.hg.1 TC0300009192.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-s9s29:80/100.66.79.146:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0