This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115617_m1
See other ABCD2 GE Assays ›
Gene Symbol:
ABCD2
Gene Name:
ATP binding cassette subfamily D member 2
Gene Aliases:
ABC39, ALDL1, ALDR, ALDRP, hALDR
Chromosome Location:
Chr.12: 39530966 - 39651333 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115617_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005164.3 NP_005155.1 4 - 5 1827 152
XM_011538027.2 XP_011536329.1 5 - 6 1767 152
XM_017018992.1 XP_016874481.1 4 - 5 1827 152
XM_017018993.1 XP_016874482.1 4 - 5 1827 152
GenBank mRNA AJ000327.1 - 4 - 5 1536 152
AK314254.1 - 4 - 5 1827 152
BC104901.1 - 4 - 5 1433 8143904 152
BC104903.1 - 4 - 5 1427 8143906 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200010413.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001392.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABCD2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
fatty acid beta-oxidation
long-chain fatty acid transport
positive regulation of fatty acid beta-oxidation
very long-chain fatty acid catabolic process
transmembrane transport
long-chain fatty acid transporter activity
protein binding
ATP binding
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12018072.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12018072.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12018072.hg.1 TC12001392.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay TC1200010413.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay PSR1200178566.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200178566.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay PSR1200178566.hg.1 ENST00000308666.3 Ensembl
Affymetrix Human Clariom D Assay PSR1200178566.hg.1 NM_005164.3 Refseq
Affymetrix Human Clariom D Assay PSR1200178566.hg.1 TC1200010413.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0