This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01115758_g1
See other GFI1 GE Assays ›
Gene Symbol:
GFI1
Gene Name:
growth factor independent 1 transcriptional repressor
Gene Aliases:
GFI-1, GFI1A, SCN2, ZNF163
Chromosome Location:
Chr.1: 92473043 - 92486876 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01115758_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GFI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127215.1 NP_001120687.1 4 - 5 954 101
NM_001127216.1 NP_001120688.1 4 - 5 947 101
NM_005263.3 NP_005254.2 4 - 5 1040 101
XM_005270749.3 XP_005270806.1 4 - 5 1396 101
XM_011541245.2 XP_011539547.1 4 - 5 1882 101
XM_011541246.2 XP_011539548.1 4 - 5 1158 101
GenBank mRNA AB209606.1 - 3 - 4 828 101
AK123009.1 - 4 - 5 1109 101
AK313876.1 - 4 - 5 954 101
BC032751.1 - 4 - 5 924 5585794 101
BC074866.2 - 3 - 4 834 30915466 101
BC074867.2 - 3 - 4 834 30915231 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100014926.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002867.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GFI1

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription involved in G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of calcidiol 1-monooxygenase activity
negative regulation of vitamin D biosynthetic process
negative regulation of neuron projection development
viral process
myeloid cell differentiation
negative regulation of NF-kappaB transcription factor activity
regulation of toll-like receptor signaling pathway
negative regulation of transcription, DNA-templated
regulation of histone H3-K4 methylation
positive regulation of interleukin-6-mediated signaling pathway
cellular response to lipopolysaccharide
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 TC01002867.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 ENST00000370332.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 ENST00000427103.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 NM_001127215.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 NM_001127216.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01044711.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay PSR0100211189.hg.1 TC0100014926.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000294702.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000370332.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000427103.5 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 ENST00000483490.1 Ensembl
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127215.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_001127216.2 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq
Affymetrix Human Clariom D Assay TC0100014926.hg.1 NM_005263.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0