Assay Details
| Gene Symbol: EPHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000120.3 | NP_000111.1 | 8 - 9 | 1421 | 80 | |
| NM_001136018.3 | NP_001129490.1 | 8 - 9 | 1273 | 80 | ||
| NM_001291163.1 | NP_001278092.1 | 8 - 9 | 1659 | 80 | ||
| GenBank mRNA | AK313436.1 | - | 8 - 9 | 1252 | 80 | |
| BC003567.1 | - | 8 - 9 | 1197 | 3547085 | 80 | |
| BC008291.1 | - | 8 - 9 | 1197 | 3604417 | 80 | |
| BC095430.1 | - | 8 - 9 | 1213 | 30377793 | 80 | |
| CR541963.1 | - | 7 - 8 | 1172 | 80 | ||
| J03518.1 | - | 8 - 9 | 1334 | 80 | ||
| KF032815.1 | - | 8 - 9 | 1381 | 80 | ||
| KU178042.1 | - | 7 - 8 | 1172 | 80 | ||
| L25878.1 | - | 8 - 9 | 1295 | 80 | ||
| L25879.1 | - | 8 - 9 | 1368 | 80 | ||
| X07936.1 | - | 8 - 9 | 1246 | 80 | ||
| Y00424.1 | - | 8 - 9 | 1324 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001856.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01028928.hg.1 | ENST00000272167.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028928.hg.1 | ENST00000366837.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028928.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028928.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028928.hg.1 | TC01001856.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000467015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000467015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000467015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000445856.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000448202.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000467015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000467015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000467015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011770.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | ENST00000272167.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | ENST00000366837.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | ENST00000614058.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | NM_000120.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | NM_001136018.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | NM_001291163.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100186887.hg.1 | TC0100011770.hg.1 | Clariom D TC |

EPHX1
Genomic Map