This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01117216_g1
See other OGG1 GE Assays ›
Gene Symbol:
OGG1
Gene Name:
8-oxoguanine DNA glycosylase
Gene Aliases:
HMMH, HOGG1, MUTM, OGH1
Chromosome Location:
Chr.3: 9749944 - 9788246 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01117216_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017006493.1 XP_016861982.1 5 - 6 1137 115
GenBank mRNA AB037879.1 - 3 - 4 281 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
base-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
response to radiation
response to light stimulus
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
response to drug
negative regulation of apoptotic process
depurination
depyrimidination
response to ethanol
response to folic acid
cellular response to cadmium ion
negative regulation of double-strand break repair via single-strand annealing
damaged DNA binding
endonuclease activity
protein binding
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
oxidized purine DNA binding
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0