This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01117250_m1
See other PRSS42 GE Assays ›
Gene Symbol:
PRSS42
Gene Name:
protease, serine 42
Gene Aliases:
TESSP2
Chromosome Location:
Chr.3: 46830404 - 46834095 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01117250_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS42
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182702.1 NP_874361.1 4 - 5 791 95
GenBank mRNA AJ544583.1 - 4 - 5 791 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300010960.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001361.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS42

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
germ cell development
spermatogenesis
serine-type endopeptidase activity

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215961 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215980 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350177 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350145 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215957 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350152 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350150 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350151 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215945 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215975 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434175 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350168 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350147 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215976 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434178 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350172 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434177 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215944 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350170 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350155 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350178 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434176 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434185 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350171 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350188 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434173 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215971 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434167 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P22350174 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434168 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P15434171 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215941 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215959 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215963 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215981 - -
Agilent Human Whole Genome ChIP Tiling Array A_17_P02215950 - -
Agilent SurePrint G3 Human CGH Microarray 4x180K 31186 - -
Agilent SurePrint G3 Human CGH Microarray 8x60K 2982 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 451259 - -
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 396288 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P408767 - -
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P408767 - -
Agilent Whole Human Genome Oligo Microarray 4x44K 41075 - -
Agilent Human Genome CGH Microarray 44B A_14_P110536 - -
Agilent Human Genome CGH Microarray 44A A_14_P110536 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 49615 - -
Agilent Human Genome CGH + SNP Microarray A_18_P14157739 - -
Agilent Human Genome CGH + SNP Microarray A_16_P16212479 - -
Agilent Human Genome CGH + SNP Microarray A_16_P16212484 - -
Agilent Human Genome CGH + SNP Microarray A_16_P16212486 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_24_P408767 - -
SurePrint G3 Human GE Microarray G4851A A_24_P408767 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350177 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350178 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215977 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434171 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215981 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434178 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215976 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434185 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350199 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434173 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434176 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215979 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350188 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434173 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215977 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350199 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434171 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215981 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350188 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215975 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434176 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215976 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434178 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350178 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P22350177 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02215979 - -
Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15434185 - -
Whole Human Genome Oligo Microarray G4112A A_24_P408767 - -
Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P110536 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215978 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215979 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215974 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215976 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215981 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215972 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215973 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215982 - -
Agilent Human Promoter ChIP-on-Chip A) A_17_P02215977 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02215976 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02215977 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434185 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434179 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434181 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434184 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434183 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434182 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434178 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P02215978 - -
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) A_17_P15434180 - -
Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P110536 - -
Agilent Human Genome CGH Microarray 4x44K A_14_P110536 - -
Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P408767 - -
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 166056 BC069619.1 genbank_mrna_human
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 166056 BC074760.2 genbank_mrna_human
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 166056 NM_002148.3 refseq_mrna_human_NM
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 166056 X59373.1 genbank_mrna_human
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 39233 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P110536 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16212474 - -
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16212486 - -
Agilent Human 1B Microarray G4111A A_32_P485345 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434181 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215954 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215952 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215966 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215981 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434177 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434185 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434175 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215962 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215976 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215959 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434171 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215972 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215943 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215977 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215964 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215968 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215960 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215945 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215950 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434173 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215938 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215974 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215957 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215979 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P15434167 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215969 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215940 - -
Agilent Human Whole Genome ChIP-on-Chip A_17_P02215948 - -
Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P110536 - -
Array Name Probe IDs Targets Target Type
Affymetrix Human Gene 1.0 ST Array NM_182702_at - -
Affymetrix Human Gene 1.0 ST Array 8086686 - -
Affymetrix Human Gene 1.0 ST Array 8086687 - -
Affymetrix Human Gene 1.0 ST Array 8086684 - -
Affymetrix Human Gene 1.0 ST Array 8086685 - -
Affymetrix Human Gene 1.0 ST Array 8086688 - -
Affymetrix Human Gene 1.1 ST Array 8086683 - -
Affymetrix Human Genome U219 Array 11742396_a_at - -
Affymetrix Human Genome U219 Array 11742396_PM_a_at - -
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03023319.hg.1 TC03001361.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03023319.hg.1 ENST00000429665.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03023319.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03023319.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay PSR0300179972.hg.1 TC0300010960.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000429665.3 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 ENST00000447340.1 Ensembl
Affymetrix Human Clariom D Assay TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay TC0300010960.hg.1 NM_182702.2 Refseq
Affymetrix Human Clariom D Assay TC0300010960.hg.1 NM_182702.2 Refseq
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A020993 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A020993 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 422268 - -
Array Name Probe IDs Targets Target Type
HumanHT-12 v4 Expression BeadChip ILMN_1757294 AJ544583.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1757294 AK092703.1 human_transcriptome
HumanHT-12 v4 Expression BeadChip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanWG-6 v2.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
HumanHT-12 V4.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_33186881-S AJ544583.1 human_transcriptome
Sentrix Human-6 Expression BeadChip GI_33186881-S NM_182702.1 human_transcriptome
Illumina HumanRef-8 v2 Expression BeadChip GI_18620-S - -
Illumina human-6 v1.0 expression beadchip 0005360563 - -
Illumina humanRef-8 v1.0 expression beadchip 0005360563 - -
Illumina human-6 v2.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina human-6 v2.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina humanRef-8 v2.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina Sentrix Human-6 v2 Expression BeadChip 510010 - -
Illumina HumanRef-6 v2.0 expression beadchip ILMN_18620 - -
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanRef-8 v3.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanWG-6 v3.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanHT-12 V3.0 expression beadchip ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanWG-6_V2_0_R2 ILMN_1757294 NM_182702.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1757294 AJ544583.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1757294 AK092703.1 human_transcriptome
Illumina HumanRef-8 WG-DASL v3.0 ILMN_1757294 NM_182702.1 human_transcriptome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0