This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01118861_m1
See other UROC1 GE Assays ›
Gene Symbol:
UROC1
Gene Name:
urocanate hydratase 1
Gene Aliases:
HMFN0320, UROCD
Chromosome Location:
Chr.3: 126481165 - 126517773 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01118861_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UROC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165974.1 NP_001159446.1 18 - 19 1943 75
NM_144639.2 NP_653240.1 17 - 18 1763 75
GenBank mRNA AB075869.1 - 18 - 19 1923 75
AK055862.1 - 17 - 18 1741 75
BC115405.1 - 17 - 18 1741 40076084 75
BC115406.1 - 17 - 18 1741 40076086 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012282.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001741.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UROC1

Panther Classification:

Molecular Function -

hydratase lyase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

histidine catabolic process
histidine catabolic process to glutamate and formamide
histidine catabolic process to glutamate and formate
urocanate hydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03031638.hg.1 TC03001741.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03031638.hg.1 ENST00000290868.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03031638.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03031638.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03031638.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay PSR0300191900.hg.1 TC0300012282.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000290868.6 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 ENST00000383579.3 Ensembl
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_001165974.1 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq
Affymetrix Human Clariom D Assay TC0300012282.hg.1 NM_144639.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0