This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01121417_g1
See other PHYKPL GE Assays ›
Gene Symbol:
PHYKPL
Gene Name:
5-phosphohydroxy-L-lysine phospho-lyase
Gene Aliases:
AGXT2L2, PHLU
Chromosome Location:
Chr.5: 178208474 - 178232822 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01121417_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHYKPL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011534669.2 XP_011532971.1 7 - 8 937 58
XM_011534670.2 XP_011532972.1 6 - 7 1270 58
XM_011534672.2 XP_011532974.1 7 - 8 723 58
XM_011534676.2 XP_011532978.1 3 - 4 877 58
XM_017009995.1 XP_016865484.1 7 - 8 937 58
XM_017009996.1 XP_016865485.1 7 - 8 937 58
XM_017009999.1 XP_016865488.1 3 - 4 2682 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

lysine catabolic process
collagen catabolic process
protein binding
transaminase activity
lyase activity
pyridoxal phosphate binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x7sxg:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0