This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01121483_m1
See other RBBP6 GE Assays ›
Gene Symbol:
RBBP6
Gene Name:
RB binding protein 6, ubiquitin ligase
Gene Aliases:
MY038, P2P-R, PACT, RBQ-1, SNAMA
Chromosome Location:
Chr.16: 24539587 - 24572863 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01121483_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBBP6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006910.4 NP_008841.2 1 - 2 1201 153
NM_018703.3 NP_061173.1 1 - 2 1201 153
NM_032626.5 NP_116015.2 1 - 2 1201 153
GenBank mRNA AB112074.1 - 1 - 2 205 153
AB112075.1 - 1 - 2 205 153
BC029352.1 - 1 - 2 292 153
BC063524.1 - 1 - 2 268 153
BC101139.2 - 1 - 2 323 153
BC101140.2 - 1 - 2 323 153
BC101141.2 - 1 - 2 323 153
BC101142.1 - 1 - 2 323 153
BC114353.1 - 1 - 2 323 153
BC114354.1 - 1 - 2 323 153
BC118667.1 - 1 - 2 323 153
BC139830.1 - 1 - 2 323 153
BX648661.1 - 1 - 2 141 153
CB529116.1 - 2 - 3 153
CD638608.1 - 1 - 2 55 153

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBBP6

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
in utero embryonic development
DNA replication
regulation of DNA replication
mRNA processing
cellular response to DNA damage stimulus
multicellular organism growth
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
embryonic organ development
somite development
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
poly(A) RNA binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0