This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01121816_m1
See other MICAL2 GE Assays ›
Gene Symbol:
MICAL2
Gene Name:
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases:
MICAL-2, MICAL2PV1, MICAL2PV2
Chromosome Location:
Chr.11: 12110576 - 12281017 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01121816_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MICAL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282664.1 NP_001269593.1 20 - 21 2933 80
XM_017018591.1 XP_016874080.1 20 - 21 2900 80
GenBank mRNA AF052170.1 - 5 - 6 626 80
AK294845.1 - 20 - 21 2900 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012966.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

heart looping
cytoskeleton organization
heart development
positive regulation of transcription via serum response element binding
sulfur oxidation
actin filament depolymerization
oxidation-reduction process
actin binding
zinc ion binding
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
NADPH:sulfur oxidoreductase activity
FAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline