This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01121819_m1
See other MICAL2 GE Assays ›
Gene Symbol:
MICAL2
Gene Name:
microtubule associated monooxygenase, calponin and LIM domain containing 2
Gene Aliases:
MICAL-2, MICAL2PV1, MICAL2PV2
Chromosome Location:
Chr.11: 12110576 - 12281017 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01121819_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MICAL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF052170.1 - 10 - 11 1272 72
AK122821.1 - 5 - 6 3758 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline