This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01122121_m1
See other ERCC8 GE Assays ›
Gene Symbol:
ERCC8
Gene Name:
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Gene Aliases:
CKN1, CSA, UVSS2
Chromosome Location:
Chr.5: 60873832 - 60945078 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01122121_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERCC8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000082.3 NP_000073.1 2 - 3 245 90
NM_001007234.2 NP_001007235.1 2 - 3 245 90
NM_001290285.1 NP_001277214.1 2 - 3 245 90
GenBank mRNA AK290726.1 - 2 - 3 225 90
AK294856.1 - 2 - 3 226 90
AK304610.1 - 2 - 3 175 90
AK314511.1 - 2 - 3 218 90
AL691658.1 - 2 - 3 232 90
BC009793.1 - 2 - 3 222 3683434 90
BT020021.1 - 2 - 3 175 90
CR536563.1 - 2 - 3 175 90
DA169067.1 - 2 - 3 245 90
KU177979.1 - 2 - 3 175 90
U28413.1 - 2 - 3 211 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK294856.1, NM_001290285.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ERCC8

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
cellular response to DNA damage stimulus
response to oxidative stress
response to UV
response to X-ray
proteasome-mediated ubiquitin-dependent protein catabolic process
protein autoubiquitination
DNA helicase activity
ubiquitin-protein transferase activity
protein binding
DNA-dependent ATPase activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05019507.hg.1 TC05001394.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05019507.hg.1 ENST00000265038.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05019507.hg.1 ENST00000426742.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05019507.hg.1 ENST00000536596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05019507.hg.1 ENST00000543101.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05019507.hg.1 NM_000082.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0