This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01125333_m1
See other PACRG GE Assays ›
Gene Symbol:
PACRG
Gene Name:
PARK2 coregulated
Gene Aliases:
GLUP, HAK005771, PARK2CRG
Chromosome Location:
Chr.6: 162726870 - 163315492 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01125333_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PACRG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080378.1 NP_001073847.1 3 - 4 519 154
NM_001080379.1 NP_001073848.1 2 - 3 561 154
NM_152410.2 NP_689623.2 3 - 4 519 154
XM_005266825.4 XP_005266882.1 2 - 3 644 154
XM_005266826.4 XP_005266883.1 2 - 3 644 154
XM_006715344.3 XP_006715407.1 2 - 3 644 154
XM_006715345.3 XP_006715408.1 2 - 3 644 154
XM_011535461.2 XP_011533763.1 2 - 3 644 154
XM_011535462.2 XP_011533764.1 2 - 3 644 154
XM_011535466.1 XP_011533768.1 2 - 3 644 154
XM_011535468.2 XP_011533770.1 2 - 3 644 154
XM_011535469.2 XP_011533771.1 2 - 3 644 154
XM_017010275.1 XP_016865764.1 3 - 4 519 154
XM_017010276.1 XP_016865765.1 3 - 4 510 154
XM_017010277.1 XP_016865766.1 3 - 4 794 154
XM_017010278.1 XP_016865767.1 3 - 4 444 154
XM_017010279.1 XP_016865768.1 3 - 4 471 154
XM_017010280.1 XP_016865769.1 2 - 3 644 154
GenBank mRNA AF546872.1 - 2 - 3 561 154
AK057286.1 - 3 - 4 519 154
BC030642.2 - 2 - 3 531 4823539 154
BC044227.1 - 3 - 4 506 5187302 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600010111.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001163.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PACRG

Gene Ontology Categories:

Function(s) Process(es)

spermatid development
cellular response to unfolded protein
negative regulation of cell death
G-protein coupled receptor binding
actin binding
Hsp70 protein binding
heat shock protein binding
ubiquitin protein ligase binding
alpha-tubulin binding
beta-tubulin binding
chaperone binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000337019.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366888.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000366889.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000534958.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000534958.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000534958.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000534958.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000534958.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000541974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542936.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542936.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000542936.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000544266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000544266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000544266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000544266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000544266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000544266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000545186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000545186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000545186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000545186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000545186.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000611387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000611387.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000621363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000621363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000621363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 ENST00000621363.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080378.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_001080379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010111.hg.1 NM_152410.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-6zkmz:80/100.66.79.198:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline