Assay Details
| Gene Symbol: CBLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284291.1 | NP_001271220.1 | 2 - 3 | 642 | 125 | |
| XM_017012644.1 | XP_016868133.1 | 2 - 3 | 836 | 125 | ||
| GenBank mRNA | BC144176.1 | - | 2 - 3 | 194 | 9052696 | 125 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012644.1 transcripts.
Set Membership: |
|

CBLL1
Genomic Map