This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01127301_g1
See other ATP5S GE Assays ›
Gene Symbol:
ATP5S
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit s (factor B)
Gene Aliases:
ATPW, FB, HSU79253
Chromosome Location:
Chr.14: 50311752 - 50326715 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01127301_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5S
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011536650.2 XP_011534952.1 4 - 5 1121 89
GenBank mRNA BG699167.1 - 4 - 5 566 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
mitochondrial ATP synthesis coupled proton transport
hydrogen ion transmembrane transporter activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000245448.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000311459.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000358473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000358473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000358473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000358473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000358473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000358473.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000426751.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000553774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000553774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000553774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000553774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000553774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000553774.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554204.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554438.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000554951.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000555939.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000556962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000556962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000556962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 ENST00000557421.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003803.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_001003805.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NM_015684.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1400007112.hg.1 NR_033761.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0