This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01128167_m1
See other EIF1AD GE Assays ›
Gene Symbol:
EIF1AD
Gene Name:
eukaryotic translation initiation factor 1A domain containing
Gene Aliases:
haponin
Chromosome Location:
Chr.11: 65996545 - 66002217 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01128167_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF1AD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001242484.1 NP_001229413.1 1 - 2 126 88
GenBank mRNA DA024526.1 - 1 - 2 120 88

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001242484.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
translation initiation factor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0