This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01360263_g1
See other PPP4R2 GE Assays ›
Gene Symbol:
PPP4R2
Gene Name:
protein phosphatase 4 regulatory subunit 2
Gene Aliases:
PP4R2
Chromosome Location:
Chr.3: 72996743 - 73069201 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01360263_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP4R2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318025.1 NP_001304954.1 1 - 2 288 93
NM_001318027.1 NP_001304956.1 1 - 2 288 93
NM_001318028.1 NP_001304957.1 1 - 2 288 93
NM_174907.3 NP_777567.1 1 - 2 288 93
GenBank mRNA AF327345.1 - 1 - 2 105 93
AJ271448.2 - 1 686 93
AK289901.1 - 1 - 2 255 93
BC100281.1 - 1 - 2 250 93
BC110889.1 - 1 - 2 245 6503859 93
BC128136.1 - 1 - 2 64 40112443 93
BC128137.1 - 1 - 2 64 40112444 93
BX089727.1 - 1 - 2 97 93
CB988831.1 - 1 - 2 297 93
DB469745.1 - 1 - 2 288 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF327345.1, AK289901.1, BC100281.1, BC110889.1, BC128136.1, BC128137.1, NM_001318025.1, NM_001318027.1, NM_001318028.1, NM_174907.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PPP4R2

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
cellular protein modification process
RNA splicing
regulation of double-strand break repair via homologous recombination
regulation of catalytic activity
protein binding
protein phosphatase type 4 regulator activity
protein binding, bridging

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500009791.hg.1 ENST00000504656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009791.hg.1 ENST00000504656.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013658.hg.1 ENST00000603917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013658.hg.1 ENST00000603917.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009988.hg.1 ENST00000429866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009988.hg.1 ENST00000429866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009995.hg.1 ENST00000411669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009995.hg.1 ENST00000411669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006518.hg.1 ENST00000512270.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006518.hg.1 ENST00000512270.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500006518.hg.1 ENST00000512270.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000476505.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000476505.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000476505.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300009995.hg.1 ENST00000411669.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009995.hg.1 ENST00000411669.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013658.hg.1 ENST00000603917.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013658.hg.1 ENST00000603917.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009791.hg.1 ENST00000504656.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009791.hg.1 ENST00000504656.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009988.hg.1 ENST00000429866.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009988.hg.1 ENST00000429866.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000356692.9 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000460360.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000470976.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000476505.2 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000476505.2 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000476505.2 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000482242.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000488810.5 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 ENST00000495566.1 Ensembl
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0300013847.hg.1 NM_174907.3 Refseq
Affymetrix Human Clariom D Assay TC0500006518.hg.1 ENST00000512270.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006518.hg.1 ENST00000512270.2 Ensembl
Affymetrix Human Clariom D Assay TC0500006518.hg.1 ENST00000512270.2 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0