This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01367736_g1
See other DISP3 GE Assays ›
Gene Symbol:
DISP3
Gene Name:
dispatched RND transporter family member 3
Gene Aliases:
PTCHD2
Chromosome Location:
Chr.1: 11479155 - 11537583 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01367736_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DISP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020780.1 NP_065831.1 5 - 6 1724 65
XM_011541828.2 XP_011540130.1 4 - 5 3826 65
XM_011541829.1 XP_011540131.1 4 - 5 699 65
XM_011541830.2 XP_011540132.1 5 - 6 1807 65
XM_011541831.2 XP_011540133.1 5 - 6 1807 65
XM_011541832.2 XP_011540134.1 5 - 6 1807 65
XM_011541833.2 XP_011540135.1 5 - 6 1807 65
XM_011541835.2 XP_011540137.1 5 - 6 1807 65
GenBank mRNA AB037758.1 - 5 - 6 1724 65
AB593117.1 - 5 - 6 1807 65
AL117236.1 - 4 - 5 644 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100006857.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000142.hg.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

smoothened signaling pathway
cholesterol metabolic process
regulation of lipid transport
cholesterol homeostasis
negative regulation of neuron differentiation
positive regulation of lipid metabolic process
positive regulation of neural precursor cell proliferation
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01002213.hg.1 TC01000142.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01002213.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01002213.hg.1 ENST00000389575.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01002213.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000423056.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000423056.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 ENST00000423056.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000304391.6 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000423056.2 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000423056.2 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 ENST00000423056.2 Ensembl
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay TC0100006857.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100147533.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100147533.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100147533.hg.1 ENST00000294484.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100147533.hg.1 NM_020780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100147533.hg.1 TC0100006857.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0