This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01370188_s1
See other GCNT2 GE Assays ›
Gene Symbol:
GCNT2
Gene Name:
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
Gene Aliases:
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1
Chromosome Location:
Chr.6: 10521283 - 10629368 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01370188_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005248997.3 XP_005249054.1 2 2665 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
protein glycosylation
transforming growth factor beta receptor signaling pathway
multicellular organism development
positive regulation of cell proliferation
posttranscriptional regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of cell-substrate adhesion
positive regulation of cell migration
positive regulation of heterotypic cell-cell adhesion
maintenance of lens transparency
positive regulation of protein kinase B signaling
positive regulation of ERK1 and ERK2 cascade
N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600010785.hg.1 ENST00000407436.1 Ensembl
Affymetrix Human Clariom D Assay TC0600010785.hg.1 ENST00000407436.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0