This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01373468_g1
See other POLR3G GE Assays ›
Gene Symbol:
POLR3G
Gene Name:
RNA polymerase III subunit G
Gene Aliases:
RPC32, RPC7
Chromosome Location:
Chr.5: 90473929 - 90514557 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01373468_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006467.2 NP_006458.2 5 - 6 556 75
XM_005248404.3 XP_005248461.1 5 - 6 521 75
XM_011543100.2 XP_011541402.1 5 - 6 789 75
XM_011543101.2 XP_011541403.1 5 - 6 448 75
XM_011543102.2 XP_011541404.1 6 - 7 678 75
XM_011543105.2 XP_011541407.1 4 - 5 629 75
XM_017008957.1 XP_016864446.1 5 - 6 1092 75
GenBank mRNA AK295434.1 - 5 - 6 521 75
BM788728.1 - 5 - 6 531 75
CD657312.1 - 2 - 3 117 75
U93868.1 - 5 - 6 518 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR3G

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
cell proliferation
positive regulation of type I interferon production
positive regulation of interferon-beta production
innate immune response
positive regulation of innate immune response
defense response to virus
RNA polymerase III activity
DNA-directed RNA polymerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000512239.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000512239.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000512239.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000399107.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503373.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000503973.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000504930.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000505345.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000512239.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000512239.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000512239.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000514483.5 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 ENST00000622335.1 Ensembl
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq
Affymetrix Human Clariom D Assay TC0500008054.hg.1 NM_006467.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0