This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01374701_g1
See other OBSCN GE Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01374701_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OBSCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 18 - 19 5487 93
NM_001271223.2 NP_001258152.2 22 - 23 6612 93
NM_052843.3 NP_443075.3 18 - 19 5487 93
XM_005273287.4 XP_005273344.1 21 - 22 6416 93
XM_005273291.4 XP_005273348.1 21 - 22 6416 93
XM_005273298.4 XP_005273355.1 21 - 22 6416 93
XM_005273307.4 XP_005273364.1 21 - 22 6416 93
XM_006711822.3 XP_006711885.1 21 - 22 6416 93
XM_006711823.3 XP_006711886.1 21 - 22 6416 93
XM_006711827.3 XP_006711890.1 21 - 22 6415 93
XM_006711829.3 XP_006711892.1 20 - 21 6139 93
XM_011544297.2 XP_011542599.1 21 - 22 6416 93
XM_011544299.2 XP_011542601.1 20 - 21 6140 93
XM_017002443.1 XP_016857932.1 21 - 22 6416 93
XM_017002444.1 XP_016857933.1 21 - 22 6379 93
XM_017002445.1 XP_016857934.1 21 - 22 6416 93
XM_017002446.1 XP_016857935.1 21 - 22 6416 93
XM_017002447.1 XP_016857936.1 21 - 22 6416 93
XM_017002448.1 XP_016857937.1 21 - 22 6266 93
XM_017002449.1 XP_016857938.1 21 - 22 6417 93
XM_017002450.1 XP_016857939.1 21 - 22 6417 93
XM_017002451.1 XP_016857940.1 21 - 22 6417 93
XM_017002452.1 XP_016857941.1 21 - 22 6417 93
XM_017002453.1 XP_016857942.1 20 - 21 6140 93
XM_017002454.1 XP_016857943.1 20 - 21 6140 93
XM_017002455.1 XP_016857944.1 20 - 21 6140 93
XM_017002456.1 XP_016857945.1 20 - 21 6140 93
XM_017002457.1 XP_016857946.1 20 - 21 6140 93
XM_017002458.1 XP_016857947.1 21 - 22 6417 93
XM_017002459.1 XP_016857948.1 21 - 22 6417 93
XM_017002460.1 XP_016857949.1 21 - 22 6417 93
XM_017002461.1 XP_016857950.1 21 - 22 6416 93
XM_017002462.1 XP_016857951.1 19 - 20 5864 93
XM_017002463.1 XP_016857952.1 19 - 20 5843 93
XM_017002464.1 XP_016857953.1 21 - 22 6416 93
XM_017002465.1 XP_016857954.1 21 - 22 6417 93
XM_017002466.1 XP_016857955.1 21 - 22 6417 93
XM_017002467.1 XP_016857956.1 21 - 22 6417 93
XM_017002468.1 XP_016857957.1 21 - 22 6417 93
XM_017002469.1 XP_016857958.1 21 - 22 6417 93
XM_017002470.1 XP_016857959.1 21 - 22 6418 93
XM_017002471.1 XP_016857960.1 21 - 22 6415 93
XM_017002472.1 XP_016857961.1 21 - 22 6415 93
XM_017002473.1 XP_016857962.1 21 - 22 6415 93
GenBank mRNA AJ002535.1 - 19 - 20 5484 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OBSCN

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0