This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01380092_m1
See other SPTY2D1 GE Assays ›
Gene Symbol:
SPTY2D1
Gene Name:
SPT2 chromatin protein domain containing 1
Gene Aliases:
Spt2
Chromosome Location:
Chr.11: 18606401 - 18634823 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01380092_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPTY2D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194285.2 NP_919261.2 3 - 4 1928 76
XM_011519919.2 XP_011518221.1 3 - 4 1786 76
GenBank mRNA AF452716.1 - 1 - 2 1322 76
AK096760.1 - 3 - 4 1948 76
AK304078.1 - 3 - 4 1844 76
BC036844.1 - 1 - 2 548 76
BX538046.1 - 1 - 2 307 76
BX641102.1 - 3 - 4 1829 76
BX647798.1 - 3 - 4 1928 76
BX648114.1 - 1 - 2 1507 76
CR749626.1 - 1 - 2 1371 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100010274.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11001470.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SPTY2D1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of chromatin assembly
histone exchange
RNA polymerase I core binding
DNA binding
histone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11018602.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018602.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018602.hg.1 TC11001470.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11018601.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11018601.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11018601.hg.1 TC11001470.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000543776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000543776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 ENST00000543776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000543776.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000543776.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 ENST00000543776.1 Ensembl
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay TC1100010274.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay PSR1100178186.hg.1 TC1100010274.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 ENST00000336349.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 ENST00000536336.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 NM_194285.2 Refseq
Affymetrix Human Clariom D Assay PSR1100178187.hg.1 TC1100010274.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline