This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01381548_m1
See other FGF13-AS1 GE Assays ›
Gene Symbol:
FGF13-AS1
Gene Name:
FGF13 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.X: 138712107 - 138716605 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01381548_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF13-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_038405.1 2 - 3 122 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008569.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000672.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Homo sapiens 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ArrayStar ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3382296 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 38973 - -
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3382296 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1560537_PM_at - -
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008814.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008814.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008814.hg.1 TC0X000672.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X008816.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008816.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008816.hg.1 TC0X000672.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X008817.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008817.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008817.hg.1 TC0X000672.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X008818.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008818.hg.1 NR_038405.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008818.hg.1 TC0X000672.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008569.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158254.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158254.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158254.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158254.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158254.hg.1 TC0X00008569.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158252.hg.1 TC0X00008569.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 ENST00000446383.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158253.hg.1 TC0X00008569.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00158250.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158250.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158250.hg.1 ENST00000438238.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00158250.hg.1 NR_038405.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00158250.hg.1 TC0X00008569.hg.1 Clariom D TC
Human Genome U133 Plus 2.0 Array 1560537_PM_at - -
Human Genome U133 Plus 2.0 Array 1560537_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at6 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1560537_at1 - -
Array Name Probe IDs Targets Target Type
Arraystar Human LncRNA microarray V2.0 ASHG19A3A041611 - -
Arraystar Human LncRNA microarray V2.0 ASHG19A3A041611 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 233144 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline