This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01385251_m1
See other APTR GE Assays ›
Gene Symbol:
APTR
Gene Name:
Alu-mediated CDKN1A/p21 transcriptional regulator (non-protein coding)
Gene Aliases:
RSBN1L-AS1
Chromosome Location:
Chr.7: 77657660 - 77697345 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01385251_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APTR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_134253.1 2 - 3 497 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000398043.2 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000416650.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000430801.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000440088.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 ENST00000447009.1 Ensembl
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq
Affymetrix Human Clariom D Assay TC0700011597.hg.1 NR_038361.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0