This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01386382_m1
See other FIRRE GE Assays ›
Gene Symbol:
FIRRE
Gene Name:
firre intergenic repeating RNA element
Gene Aliases:
LINC01200
Chromosome Location:
Chr.X: 131702650 - 131830643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01386382_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FIRRE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_026975.1 1 - 2 352 101
GenBank mRNA BC131554.1 - 1 - 2 146 101
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB108042.1 - 1 - 2 352 Chr. X - 131804962 - 131830643 [-] on Build GRCh38 101
GenBankmRNA DB129133.1 - 1 - 2 234 Chr. X - 131794350 - 131830525 [-] on Build GRCh38 101
GenBankmRNA BE386147.1 - 1 - 2 283 Chr. X - 131796240 - 131830574 [-] on Build GRCh38 101
GenBankmRNA BF904499.1 - 3 - 4 Chr. X - 131796289 - 131830420 [+] on Build GRCh38 101
GenBankmRNA CN371302.1 - 1 - 2 299 Chr. X - 131804960 - 131830575 [-] on Build GRCh38 101
GenBankmRNA DA374979.1 - 1 - 2 336 Chr. X - 131804962 - 131830627 [-] on Build GRCh38 101
GenBankmRNA DB106734.1 - 234 Chr. - - [-] on Build 101
GenBankmRNA BE257517.1 - 1 - 2 326 Chr. X - 131794369 - 131830617 [-] on Build GRCh38 101
GenBankmRNA BX399893.2 - 1 - 2 298 Chr. X - 131795869 - 131830589 [-] on Build GRCh38 101
GenBankmRNA BE385351.1 - 1 - 2 283 Chr. X - 131796316 - 131830574 [-] on Build GRCh38 101
GenBankmRNA BE409638.1 - 1 - 2 315 Chr. X - 131796124 - 131830578 [-] on Build GRCh38 101
GenBankmRNA BF572469.1 - 1 - 2 333 Chr. X - 131824314 - 131830623 [-] on Build GRCh38 101
GenBankmRNA BI906044.1 - 1 - 2 322 Chr. X - 131795957 - 131830613 [-] on Build GRCh38 101

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream olfactory receptor family 13 subfamily H member 1 OR13H1 ORX1 -280.2kb Anti-sense Chr. X - 131544074 - 131545000 on NCBI Build GRCh38
Downstream serine/threonine protein kinase 26 STK26 MASK,
MST4
192.6kb Anti-sense Chr. X - 132023217 - 132075943 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00010826.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001366.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000394346.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000394346.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000394346.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000394346.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000394346.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 ENST00000427391.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010826.hg.1 NR_026975.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0