This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01390879_m1
See other CDKN2B-AS1 GE Assays ›
Gene Symbol:
CDKN2B-AS1
Gene Name:
CDKN2B antisense RNA 1
Gene Aliases:
ANRIL, CDKN2B-AS, CDKN2BAS, NCRNA00089, PCAT12, p15AS
Chromosome Location:
Chr.9: 21994791 - 22121097 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01390879_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKN2B-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003529.3 1 - 2 371 133
NR_047532.1 1 - 2 371 133
NR_047539.1 1 - 2 371 133
GenBank mRNA AB548314.1 - 1 - 2 371 133
BI860238.1 - 2 - 3 186 133
CB109081.1 - 1 - 2 371 133
DQ485453.1 - 1 - 2 370 133
DQ485454.1 - 1 - 2 370 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0