This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01391550_m1
See other PTPRD-AS1 GE Assays ›
Gene Symbol:
PTPRD-AS1
Gene Name:
PTPRD antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 8858018 - 8861727 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01391550_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRD-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_121600.1 1 - 2 309 106
GenBank mRNA DB495850.1 - 1 - 2 185 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900006622.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121600.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121600.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900006622.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145327.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145327.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145327.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145327.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145327.hg.1 TC0900006622.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145329.hg.1 TC0900006622.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay PSR0900145332.hg.1 TC0900006622.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000430766.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 ENST00000447950.1 Ensembl
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121599.1 Refseq
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121600.1 Refseq
Affymetrix Human Clariom D Assay TC0900006622.hg.1 NR_121600.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0