This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01396878_m1
See other PLCH1-AS1 GE Assays ›
Gene Symbol:
PLCH1-AS1
Gene Name:
PLCH1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 155449185 - 155457753 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01396878_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLCH1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC039489.2 - 1 - 2 244 128
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM472067.1 - 1 - 2 256 Chr. 3 - 155449182 - 155457491 [+] on Build GRCh38 128

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC100507537 LOC100507537 -155.5kb Anti-sense Chr. 3 - 155290232 - 155293700 on NCBI Build GRCh38
Downstream phospholipase C eta 1 PLCH1 PLCL3 22.4kb Anti-sense Chr. 3 - 155458603 - 155745723 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300009261.hg.1 ENST00000475196.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300009261.hg.1 ENST00000475196.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009261.hg.1 ENST00000475196.1 Ensembl
Affymetrix Human Clariom D Assay TC0300009261.hg.1 ENST00000475196.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0