This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01397577_m1
See other DSEL GE Assays ›
Gene Symbol:
DSEL
Gene Name:
dermatan sulfate epimerase-like
Gene Aliases:
C18orf4
Chromosome Location:
Chr.18: 67506582 - 67516730 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01397577_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DSEL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032160.2 NP_115536.1 1 - 2 370 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800008952.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000564.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_032160.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DSEL

Gene Ontology Categories:

Function(s) Process(es)

chondroitin sulfate metabolic process
dermatan sulfate biosynthetic process
sulfotransferase activity
chondroitin-glucuronate 5-epimerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR18005875.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005875.hg.1 ENST00000397964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005875.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005875.hg.1 TC18000564.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1800007583.hg.1 ENST00000581951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007583.hg.1 ENST00000581951.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007583.hg.1 ENST00000583493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007583.hg.1 ENST00000583493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007583.hg.1 ENST00000583493.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005881.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005881.hg.1 ENST00000397964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005881.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005881.hg.1 TC18000564.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR18005878.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005878.hg.1 ENST00000397964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005878.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005878.hg.1 TC18000564.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR18005879.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005879.hg.1 ENST00000397964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005879.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005879.hg.1 TC18000564.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005877.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005877.hg.1 ENST00000397964.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR18005877.hg.1 NM_032160.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR18005877.hg.1 TC18000564.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1800161420.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161420.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161420.hg.1 TC1800008952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800161420.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161420.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161419.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161419.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161419.hg.1 TC1800008952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800161419.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161419.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161421.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161421.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161421.hg.1 TC1800008952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800161421.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161421.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800008952.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161422.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161422.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay PSR1800161422.hg.1 TC1800008952.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800161422.hg.1 ENST00000310045.7 Ensembl
Affymetrix Human Clariom D Assay PSR1800161422.hg.1 NM_032160.2 Refseq
Affymetrix Human Clariom D Assay TC1800007583.hg.1 ENST00000581951.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007583.hg.1 ENST00000581951.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007583.hg.1 ENST00000583493.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007583.hg.1 ENST00000583493.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007583.hg.1 ENST00000583493.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0