This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01399300_m1
See other PRSS37 GE Assays ›
Gene Symbol:
PRSS37
Gene Name:
protease, serine 37
Gene Aliases:
TRYX2
Chromosome Location:
Chr.7: 141836278 - 141849725 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01399300_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS37
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008270.2 NP_001008271.2 1 - 2 409 97
NM_001171951.1 NP_001165422.1 1 - 2 409 97
XM_005249946.4 XP_005250003.1 1 - 2 8713 97
XM_005249947.3 XP_005250004.1 2 - 3 1531 97
GenBank mRNA AK097648.1 - 2 - 3 89 97
AK172866.1 - 1 - 2 409 97
BC137353.1 - 1 - 2 199 9021351 97
BC137354.1 - 1 - 2 199 9021352 97
BC144605.1 - 1 - 2 199 9053136 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012847.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001933.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC137353.1,BC137354.1,BC144605.1,NM_001008270.2,NM_001171951.1,XM_005249946.4,XM_005249947.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS37

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
binding of sperm to zona pellucida
cell migration
protein maturation
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07030044.hg.1 TC07001933.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07030044.hg.1 ENST00000350549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030044.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030044.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030044.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030046.hg.1 TC07001933.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07030046.hg.1 ENST00000350549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030046.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030046.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030045.hg.1 TC07001933.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07030045.hg.1 ENST00000350549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030045.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030045.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030042.hg.1 TC07001933.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07030042.hg.1 ENST00000350549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030042.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030042.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030042.hg.1 NM_001171951.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030043.hg.1 TC07001933.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07030043.hg.1 ENST00000350549.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030043.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07030043.hg.1 NM_001008270.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07030043.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192616.hg.1 TC0700012847.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192615.hg.1 TC0700012847.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192612.hg.1 TC0700012847.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192613.hg.1 TC0700012847.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay TC0700012847.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000350549.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000419085.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000438520.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 ENST00000452758.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 NM_001008270.2 Refseq
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 NM_001171951.1 Refseq
Affymetrix Human Clariom D Assay PSR0700192611.hg.1 TC0700012847.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 200353 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0