This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01546597_m1
See other GOT1 GE Assays ›
Gene Symbol:
GOT1
Gene Name:
glutamic-oxaloacetic transaminase 1
Gene Aliases:
AST1, ASTQTL1, GIG18, cAspAT, cCAT
Chromosome Location:
Chr.10: 99396870 - 99430773 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01546597_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GOT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002079.2 NP_002070.1 7 - 8 1168 62
GenBank mRNA AF052153.1 - 7 - 8 988 62
AK293483.1 - 7 - 8 890 62
AK301916.1 - 7 - 8 1066 62
AK312684.1 - 7 - 8 1010 62
AY513279.1 - 7 - 8 962 62
BC000498.2 - 7 - 8 988 2820849 62
M37400.1 - 7 - 8 984 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011585.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001580.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GOT1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
2-oxoglutarate metabolic process
oxaloacetate metabolic process
glycerol biosynthetic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
glutamate metabolic process
Notch signaling pathway
cellular amino acid biosynthetic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
cellular response to insulin stimulus
response to glucocorticoid
fatty acid homeostasis
L-aspartate:2-oxoglutarate aminotransferase activity
phosphatidylserine decarboxylase activity
pyridoxal phosphate binding
carboxylic acid binding
identical protein binding
L-cysteine:2-oxoglutarate aminotransferase activity
L-phenylalanine:2-oxoglutarate aminotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10020303.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10020303.hg.1 ENST00000535447.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10020303.hg.1 ENST00000543866.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10020303.hg.1 NM_002079.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10020303.hg.1 TC10001580.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000471741.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay TC1000011585.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 ENST00000370508.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 ENST00000489349.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 NM_002079.2 Refseq
Affymetrix Human Clariom D Assay PSR1000178976.hg.1 TC1000011585.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0