This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547266_m1
See other ZC3H14 GE Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547266_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZC3H14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160103.1 NP_001153575.1 9 - 10 1501 129
NM_001160104.1 NP_001153576.1 9 - 10 1501 129
NM_001326295.1 NP_001313224.1 9 - 10 1501 129
NM_001326297.1 NP_001313226.1 9 - 10 1682 129
NM_001326300.1 NP_001313229.1 8 - 9 1386 129
NM_001326302.1 NP_001313231.1 8 - 9 1386 129
NM_001326304.1 NP_001313233.1 8 - 9 1386 129
NM_001326308.1 NP_001313237.1 8 - 9 1386 129
NM_001326310.1 NP_001313239.1 9 - 10 1501 129
NM_001326315.1 NP_001313244.1 9 - 10 1682 129
NM_001326316.1 NP_001313245.1 9 - 10 1349 129
NM_024824.4 NP_079100.2 9 - 10 1501 129
NM_207661.2 NP_997544.1 9 - 10 1349 129
NR_136936.1 9 - 10 1501 129
XM_005268067.4 XP_005268124.1 9 - 10 1496 129
XM_005268069.4 XP_005268126.1 9 - 10 1496 129
XM_005268070.4 XP_005268127.1 9 - 10 1496 129
XM_011537160.2 XP_011535462.1 9 - 10 1609 129
GenBank mRNA AK303123.1 - 10 - 11 1345 129
AK304275.1 - 9 - 10 1372 129
AL834215.1 - 9 - 10 1349 129
AY578060.1 - 9 - 10 1385 129
AY578062.1 - 9 - 10 1349 129
AY578063.1 - 2 - 3 223 129
BC011793.1 - 9 - 10 1367 4298961 129
BC023641.2 - 5 - 6 1007 129
BX248265.1 - 5 - 6 1023 129
BX640716.1 - 9 - 10 1380 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1400007906.hg.1
This assay targets the same location as HTA transcript cluster(s) TC14000536.hg.1

Additional Information:

This assay is in the 5'UTR region of the AY578063.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZC3H14

Gene Ontology Categories:

Function(s) Process(es)

regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000251038.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000318308.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000336693.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000353091.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000359301.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000380684.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000555755.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000555900.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 ENST00000556945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 NM_001160103.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 NM_001160104.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 NM_024824.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 NM_207661.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 NM_207662.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR14006600.hg.1 TC14000536.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000251038.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000318308.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000336693.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000555755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000555900.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000556000.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000556945.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_001160103.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_001160104.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_024824.4 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_207661.2 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_207662.3 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000251038.9 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000318308.10 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000336693.8 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000555755.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000555900.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000556000.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 ENST00000556945.5 Ensembl
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_001160103.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_001160104.1 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_024824.4 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_207661.2 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 NM_207662.3 Refseq
Affymetrix Human Clariom D Assay PSR1400156024.hg.1 TC1400007906.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0