This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547572_g1
See other CTH GE Assays ›
Gene Symbol:
CTH
Gene Name:
cystathionine gamma-lyase
Gene Aliases:
-
Chromosome Location:
Chr.1: 70411218 - 70441949 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547572_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190463.1 NP_001177392.1 5 - 6 746 99
NM_001902.5 NP_001893.2 6 - 7 842 99
XM_005270509.3 XP_005270566.1 6 - 7 862 99
XM_017000416.1 XP_016855905.1 3 - 4 566 99
GenBank mRNA AK223376.1 - 6 - 7 815 99
AK303946.1 - 5 - 6 723 99
AK310269.1 - 7 - 8 888 99
BC015807.2 - 6 - 7 788 3915913 99
BT006882.1 - 6 - 7 644 99
S52784.1 - 6 - 7 677 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CTH

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

sulfur amino acid catabolic process
cysteine metabolic process
protein-pyridoxal-5-phosphate linkage via peptidyl-N6-pyridoxal phosphate-L-lysine
cysteine biosynthetic process via cystathionine
cysteine biosynthetic process
transsulfuration
endoplasmic reticulum unfolded protein response
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein sulfhydration
positive regulation of NF-kappaB transcription factor activity
protein homotetramerization
hydrogen sulfide biosynthetic process
'de novo' L-methionine biosynthetic process
positive regulation of aortic smooth muscle cell differentiation
negative regulation of apoptotic signaling pathway
cystathionine gamma-synthase activity
cystathionine gamma-lyase activity
protein binding
calmodulin binding
carbon-sulfur lyase activity
pyridoxal phosphate binding
L-cystine L-cysteine-lyase (deaminating)
homocysteine desulfhydrase activity
L-cysteine desulfhydrase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline