This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547876_g1
See other HACD1 GE Assays ›
Gene Symbol:
HACD1
Gene Name:
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases:
CAP, PTPLA
Chromosome Location:
Chr.10: 17589959 - 17617374 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547876_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014241.3 NP_055056.3 2 - 3 407 130
GenBank mRNA BC010353.1 - 2 - 3 411 4293342 130

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HACD1

Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
multicellular organism development
long-chain fatty-acyl-CoA biosynthetic process
3R-hydroxyacyl-CoA dehydratase activity
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0