This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01547879_m1
See other HACD1 GE Assays ›
Gene Symbol:
HACD1
Gene Name:
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases:
CAP, PTPLA
Chromosome Location:
Chr.10: 17589959 - 17617374 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01547879_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014241.3 NP_055056.3 5 - 6 640 78
XM_005252641.4 XP_005252698.1 3 - 4 531 78
GenBank mRNA AF114494.1 - 5 - 6 653 78
BC010353.1 - 5 - 6 644 4293342 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000009930.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001075.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HACD1

Gene Ontology Categories:

Function(s) Process(es)

fatty acid biosynthetic process
multicellular organism development
long-chain fatty-acyl-CoA biosynthetic process
3R-hydroxyacyl-CoA dehydratase activity
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10013606.hg.1 ENST00000361271.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013606.hg.1 NM_014241.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10013606.hg.1 TC10001075.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000326961.6 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000466335.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 ENST00000632169.1 Ensembl
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay TC1000009930.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 ENST00000361271.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 ENST00000471481.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 ENST00000498812.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 NM_014241.3 Refseq
Affymetrix Human Clariom D Assay PSR1000167889.hg.1 TC1000009930.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0