This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01548162_m1
See other RAMP1 GE Assays ›
Gene Symbol:
RAMP1
Gene Name:
receptor activity modifying protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 237858714 - 237912117 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01548162_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAMP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308353.1 NP_001295282.1 3 - 4 384 85
NM_005855.3 NP_005846.1 2 - 3 323 85
XM_017003152.1 XP_016858641.1 2 - 3 245 85
XM_017003153.1 XP_016858642.1 2 - 3 553 85
XM_017003154.1 XP_016858643.1 2 - 3 330 85
XM_017003156.1 XP_016858645.1 2 - 3 271 85
GenBank mRNA AI017108.1 - 2 - 3 323 85
AJ001014.1 - 2 - 3 223 85
AY265457.1 - 2 - 3 191 85
BC000548.2 - 2 - 3 215 3163522 85
BI548986.1 - 3 - 4 310 85
BM978161.1 - 1 - 2 603 85
CR542032.1 - 2 - 3 191 85
CR542044.1 - 2 - 3 191 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAMP1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
calcium ion transport
intracellular protein transport
G-protein coupled receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
positive regulation of protein glycosylation
protein localization to plasma membrane
receptor activity
calcitonin receptor activity
protein binding
protein transporter activity
coreceptor activity
calcitonin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0