This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01548230_g1
See other HADHA GE Assays ›
Gene Symbol:
HADHA
Gene Name:
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit
Gene Aliases:
ECHA, GBP, HADH, LCEH, LCHAD, MTPA, TP-ALPHA
Chromosome Location:
Chr.2: 26190635 - 26244726 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01548230_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HADHA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000182.4 NP_000173.2 6 - 7 707 67
GenBank mRNA AK293398.1 - 3 - 4 200 67
AK302532.1 - 5 - 6 454 67
AK313027.1 - 6 - 7 600 67
BC009235.2 - 6 - 7 602 2966432 67
D16480.1 - 6 - 7 604 67
FJ626228.1 - 5 - 6 443 67
GU727642.1 - 6 - 7 602 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200012030.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001665.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK293398.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HADHA

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
response to insulin
cardiolipin acyl-chain remodeling
response to drug
fatty-acyl-CoA binding
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acetyltransferase activity
enoyl-CoA hydratase activity
protein binding
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
protein complex binding
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02026672.hg.1 TC02001665.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026672.hg.1 ENST00000380649.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026672.hg.1 ENST00000457468.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026672.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02026673.hg.1 TC02001665.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02026673.hg.1 ENST00000380649.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026673.hg.1 ENST00000457468.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02026673.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay PSR0200187546.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200187546.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay PSR0200187546.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200187546.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay PSR0200187546.hg.1 TC0200012030.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200187545.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200187545.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay PSR0200187545.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay PSR0200187545.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay PSR0200187545.hg.1 TC0200012030.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000380649.7 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000461025.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000471743.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 ENST00000492433.1 Ensembl
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq
Affymetrix Human Clariom D Assay TC0200012030.hg.1 NM_000182.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0