Assay Details
| Gene Symbol: GPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142466.2 | NP_001135938.1 | 8 - 9 | 1121 | 64 | |
| NM_133443.3 | NP_597700.1 | 8 - 9 | 1145 | 64 | ||
| XM_017023790.1 | XP_016879279.1 | 5 - 6 | 618 | 64 | ||
| GenBank mRNA | AK094971.1 | - | 8 - 9 | 1045 | 64 | |
| AY029173.2 | - | 8 - 9 | 1129 | 64 | ||
| BC062555.1 | - | 8 - 9 | 1094 | 6429257 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | TC1600007723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156359.hg.1 | NM_133443.3 | Refseq |

GPT2
Genomic Map