This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01548679_g1
See other GPT GE Assays ›
Gene Symbol:
GPT
Gene Name:
glutamic--pyruvic transaminase
Gene Aliases:
AAT1, ALT1, GPT1
Chromosome Location:
Chr.8: 144502217 - 144507174 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01548679_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005309.2 NP_005300.1 9 - 10 1513 99
XM_011516993.2 XP_011515295.1 10 - 11 2203 99
GenBank mRNA BC018207.1 - 10 - 11 1447 4179699 99
BT006992.1 - 9 - 10 1290 99
D10355.1 - 9 - 10 1281 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800009265.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08000856.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPT

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid biosynthetic process
L-alanine catabolic process
L-alanine:2-oxoglutarate aminotransferase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08011446.hg.1 TC08000856.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08011446.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08011446.hg.1 ENST00000528431.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08011446.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay PSR0800163561.hg.1 TC0800009265.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000354769.8 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000394955.2 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527165.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000527961.1 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000528431.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000531330.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 ENST00000534702.5 Ensembl
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq
Affymetrix Human Clariom D Assay TC0800009265.hg.1 NM_005309.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0