This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01548685_m1
See other ARHGAP39 GE Assays ›
Gene Symbol:
ARHGAP39
Gene Name:
Rho GTPase activating protein 39
Gene Aliases:
CrGAP, Vilse
Chromosome Location:
Chr.8: 144529179 - 144700520 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01548685_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308207.1 NP_001295136.1 7 - 8 3069 73
NM_001308208.1 NP_001295137.1 6 - 7 2722 73
GenBank mRNA AB051475.1 - 7 - 8 3073 73
AK296353.1 - 6 - 7 2719 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800012225.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay PSR0800183993.hg.1 TC0800012225.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0