This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01548687_m1
See other ARHGAP39 GE Assays ›
Gene Symbol:
ARHGAP39
Gene Name:
Rho GTPase activating protein 39
Gene Aliases:
CrGAP, Vilse
Chromosome Location:
Chr.8: 144529179 - 144700520 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01548687_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP39
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308207.1 NP_001295136.1 9 - 10 3352 66
NM_001308208.1 NP_001295137.1 8 - 9 3005 66
NM_025251.2 NP_079527.1 8 - 9 2971 66
XM_011517308.1 XP_011515610.1 9 - 10 3252 66
XM_011517309.1 XP_011515611.1 9 - 10 3156 66
XM_011517312.2 XP_011515614.1 7 - 8 3790 66
XM_017013870.1 XP_016869359.1 10 - 11 4324 66
XM_017013871.1 XP_016869360.1 9 - 10 3383 66
GenBank mRNA AB051475.1 - 9 - 10 3356 66
AK092501.1 - 4 - 5 418 66
AK296353.1 - 8 - 9 3002 66
AK303851.1 - 5 - 6 1674 66
BC013071.2 - 2 - 3 282 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800012225.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGAP39

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay PSR0800183991.hg.1 TC0800012225.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000276826.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000377307.6 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 ENST00000528810.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308207.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_001308208.1 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq
Affymetrix Human Clariom D Assay TC0800012225.hg.1 NM_025251.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0